3-(Benzyloxy)-N-(2,4-dichlorobenzyl)aniline

Supplier: Matrix Scientific

032142-500MG
101833-108EA 306.15 USD
101833-108
3-(Benzyloxy)-N-(2,4-dichlorobenzyl)aniline
3-(Benzyloxy)-N-(2,4-dichlorobenzyl)aniline

Hazard: Irritant

Formula: C₂₀H₁₇Cl₂NO
MDL Number: MFCD10688148
CAS Number: 1040688-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR