3-(Benzyloxy)-N-[2-(2-isopropylphenoxy)ethyl]aniline

Supplier: Matrix Scientific

032140-500MG
101833-104EA 306.15 USD
101833-104
3-(Benzyloxy)-N-[2-(2-isopropylphenoxy)ethyl]aniline
3-(Benzyloxy)-N-[2-(2-isopropylphenoxy)ethyl]aniline

Hazard: Irritant

Formula: C₂₄H₂₇NO₂
MW: 361.49 g/mol
MDL Number: MFCD10688146
CAS Number: 1040688-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR