N-[4-(2-Methoxyethoxy)benzyl]-2-(3-phenylpropoxy)aniline

Supplier: Matrix Scientific

032138-500MG
101833-100EA 306.15 USD
101833-100
N-[4-(2-Methoxyethoxy)benzyl]-2-(3-phenylpropoxy)aniline
N-[4-(2-Methoxyethoxy)benzyl]-2-(3-phenylpropoxy)aniline

Hazard: Irritant

Formula: C₂₅H₂₉NO₃
MDL Number: MFCD10688144
CAS Number: 1040688-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR