N-Phenethyl-2-(3-phenylpropoxy)aniline

Supplier: Matrix Scientific

032132-500MG
101833-088EA 306.15 USD
101833-088
N-Phenethyl-2-(3-phenylpropoxy)aniline
N-Phenethyl-2-(3-phenylpropoxy)aniline

Hazard: Irritant

Formula: C₂₃H₂₅NO
MDL Number: MFCD10688138
CAS Number: 1040687-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR