N-(2-Phenoxyethyl)-2-(3-phenylpropoxy)aniline

Supplier: Matrix Scientific

032130-500MG
101833-084EA 306.15 USD
101833-084
N-(2-Phenoxyethyl)-2-(3-phenylpropoxy)aniline
N-(2-Phenoxyethyl)-2-(3-phenylpropoxy)aniline

Hazard: Irritant

Formula: C₂₃H₂₅NO₂
MDL Number: MFCD10688136
CAS Number: 1040687-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR