N-[2-(2,6-Dimethylphenoxy)ethyl]-2-(3-phenylpropoxy)aniline

Supplier: Matrix Scientific

032128-500MG
101833-080EA 306.15 USD
101833-080
N-[2-(2,6-Dimethylphenoxy)ethyl]-2-(3-phenylpropoxy)aniline
N-[2-(2,6-Dimethylphenoxy)ethyl]-2-(3-phenylpropoxy)aniline

Hazard: Irritant

Formula: C₂₅H₂₉NO₂
MDL Number: MFCD10688134
CAS Number: 1040687-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR