3,4-Dimethyl-N-[2-(4-methylphenoxy)propyl]aniline

Supplier: Matrix Scientific

032127-500MG
101833-078EA 306.15 USD
101833-078
3,4-Dimethyl-N-[2-(4-methylphenoxy)propyl]aniline
3,4-Dimethyl-N-[2-(4-methylphenoxy)propyl]aniline

Hazard: Irritant

Formula: C₁₈H₂₃NO
MDL Number: MFCD10688133
CAS Number: 1040687-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR