N-[4-(sec-Butoxy)benzyl]-4-(2-ethoxyethoxy)aniline

Supplier: Matrix Scientific

032116-500MG
101833-056EA 306.15 USD
101833-056
N-[4-(sec-Butoxy)benzyl]-4-(2-ethoxyethoxy)aniline
N-[4-(sec-Butoxy)benzyl]-4-(2-ethoxyethoxy)aniline

Hazard: Irritant

Formula: C₂₁H₂₉NO₃
MDL Number: MFCD10688122
CAS Number: 1040687-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR