N-(3,5-Dichlorobenzyl)-4-(2-ethoxyethoxy)aniline

Supplier: Matrix Scientific

032115-500MG
101833-054EA 306.15 USD
101833-054
N-(3,5-Dichlorobenzyl)-4-(2-ethoxyethoxy)aniline
N-(3,5-Dichlorobenzyl)-4-(2-ethoxyethoxy)aniline

Hazard: Irritant

Formula: C₁₇H₁₉Cl₂NO₂
MDL Number: MFCD10688121
CAS Number: 1040687-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR