N-(3-Ethoxybenzyl)-4-(2-ethoxyethoxy)aniline
Supplier: Matrix Scientific
032112-500MG
101833-048EA
306.15
USD
101833-048
N-(3-Ethoxybenzyl)-4-(2-ethoxyethoxy)aniline
N-(3-Ethoxybenzyl)-4-(2-ethoxyethoxy)aniline
Hazard: Irritant
Formula:
C₁₉H₂₅NO₃ |
MDL Number:
MFCD10688118 CAS Number: 1040687-12-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...