N-[2-(4-Chloro-3-methylphenoxy)ethyl]-4-(2-ethoxyethoxy)aniline

Supplier: Matrix Scientific

032107-500MG
101833-038EA 306.15 USD
101833-038
N-[2-(4-Chloro-3-methylphenoxy)ethyl]-4-(2-ethoxyethoxy)aniline
N-[2-(4-Chloro-3-methylphenoxy)ethyl]-4-(2-ethoxyethoxy)aniline

Hazard: Irritant

Formula: C₁₉H₂₄ClNO₃
MDL Number: MFCD10688115
CAS Number: 1040686-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR