N-[2-(Phenethyloxy)benzyl]-4-(3-phenylpropoxy)aniline

Supplier: Matrix Scientific

032106-500MG
101833-036EA 306.15 USD
101833-036
N-[2-(Phenethyloxy)benzyl]-4-(3-phenylpropoxy)aniline
N-[2-(Phenethyloxy)benzyl]-4-(3-phenylpropoxy)aniline

Hazard: Irritant

Formula: C₃₀H₃₁NO₂
MW: 437.59 g/mol
MDL Number: MFCD10688114
CAS Number: 1040686-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR