N-[4-(Heptyloxy)benzyl]-4-(3-phenylpropoxy)aniline

Supplier: Matrix Scientific

032105-500MG
101833-034EA 306.15 USD
101833-034
N-[4-(Heptyloxy)benzyl]-4-(3-phenylpropoxy)aniline
N-[4-(Heptyloxy)benzyl]-4-(3-phenylpropoxy)aniline

Hazard: Irritant

Formula: C₂₉H₃₇NO₂
MDL Number: MFCD10688113
CAS Number: 1040686-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR