N-[2-(2,4-Dichlorophenoxy)ethyl]-3,4-dimethylaniline

Supplier: Matrix Scientific

032103-500MG
101833-030EA 306.15 USD
101833-030
N-[2-(2,4-Dichlorophenoxy)ethyl]-3,4-dimethylaniline
N-[2-(2,4-Dichlorophenoxy)ethyl]-3,4-dimethylaniline

Hazard: Irritant

Formula: C₁₆H₁₇Cl₂NO
MDL Number: MFCD03210950
CAS Number: 356532-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR