N-(4-Chlorophenethyl)-4-(3-phenylpropoxy)aniline

Supplier: Matrix Scientific

032102-500MG
101833-028EA 306.15 USD
101833-028
N-(4-Chlorophenethyl)-4-(3-phenylpropoxy)aniline
N-(4-Chlorophenethyl)-4-(3-phenylpropoxy)aniline

Hazard: Irritant

Formula: C₂₃H₂₄ClNO
MDL Number: MFCD10688111
CAS Number: 1040686-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR