N-[4-(2,4-Dichlorophenoxy)butyl]-4-(3-phenylpropoxy)aniline

Supplier: Matrix Scientific

032101-500MG
101833-026EA 306.15 USD
101833-026
N-[4-(2,4-Dichlorophenoxy)butyl]-4-(3-phenylpropoxy)aniline
N-[4-(2,4-Dichlorophenoxy)butyl]-4-(3-phenylpropoxy)aniline

Hazard: Irritant

Formula: C₂₅H₂₇Cl₂NO₂
MDL Number: MFCD10688110
CAS Number: 1040686-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR