3,4-Dimethyl-N-(3-methylbenzyl)aniline

Supplier: Matrix Scientific

032099-500MG
101833-022EA 306.15 USD
101833-022
3,4-Dimethyl-N-(3-methylbenzyl)aniline
3,4-Dimethyl-N-(3-methylbenzyl)aniline

Hazard: Irritant

Formula: C₁₆H₁₉N
MDL Number: MFCD03210614
CAS Number: 356530-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR