N-[2-(3,5-Dimethylphenoxy)ethyl]-4-(3-phenylpropoxy)aniline

Supplier: Matrix Scientific

032098-500MG
101833-020EA 306.15 USD
101833-020
N-[2-(3,5-Dimethylphenoxy)ethyl]-4-(3-phenylpropoxy)aniline
N-[2-(3,5-Dimethylphenoxy)ethyl]-4-(3-phenylpropoxy)aniline

Hazard: Irritant

Formula: C₂₅H₂₉NO₂
MDL Number: MFCD10688108
CAS Number: 1040686-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR