N-[4-(2-Ethoxyethoxy)benzyl]-3-(2-phenoxyethoxy)aniline

Supplier: Matrix Scientific

032096-500MG
101833-016EA 306.15 USD
101833-016
N-[4-(2-Ethoxyethoxy)benzyl]-3-(2-phenoxyethoxy)aniline
N-[4-(2-Ethoxyethoxy)benzyl]-3-(2-phenoxyethoxy)aniline

Hazard: Irritant

Formula: C₂₅H₂₉NO₄
MDL Number: MFCD10688106
CAS Number: 1040686-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR