N-[2-(3-Methylphenoxy)propyl]-3-(2-phenoxyethoxy)aniline

Supplier: Matrix Scientific

032092-500MG
101833-008EA 306.15 USD
101833-008
N-[2-(3-Methylphenoxy)propyl]-3-(2-phenoxyethoxy)aniline
N-[2-(3-Methylphenoxy)propyl]-3-(2-phenoxyethoxy)aniline

Hazard: Irritant

Formula: C₂₄H₂₇NO₃
MDL Number: MFCD10688103
CAS Number: 1040686-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR