N-Neopentyl-3-(2-phenoxyethoxy)aniline

Supplier: Matrix Scientific

032091-500MG
101833-006EA 306.15 USD
101833-006
N-Neopentyl-3-(2-phenoxyethoxy)aniline
N-Neopentyl-3-(2-phenoxyethoxy)aniline

Hazard: Irritant

Formula: C₁₉H₂₅NO₂
MDL Number: MFCD10688102
CAS Number: 1040686-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR