N-[4-(tert-Butyl)benzyl]-3-(2-phenoxyethoxy)-aniline

Supplier: Matrix Scientific

032089-500MG
101833-002EA 260.23 USD
101833-002
N-[4-(tert-Butyl)benzyl]-3-(2-phenoxyethoxy)-aniline
N-[4-(tert-Butyl)benzyl]-3-(2-phenoxyethoxy)aniline

CAS: 1040686-14-3
MDL Number: MFCD10688100
Molecular Formula: C25H29NO2
Molecular Weight: 375,52
Hazard: Irritant

Formula: C₂₅H₂₉NO₂
MDL Number: MFCD10688100
CAS Number: 1040686-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR