3-(2-Phenoxyethoxy)-N-(3-phenylpropyl)aniline

Supplier: Matrix Scientific

032087-500MG
101832-998EA 306.15 USD
101832-998
3-(2-Phenoxyethoxy)-N-(3-phenylpropyl)aniline
3-(2-Phenoxyethoxy)-N-(3-phenylpropyl)aniline

Hazard: Irritant

Formula: C₂₃H₂₅NO₂
MDL Number: MFCD10688098
CAS Number: 1040686-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR