N-([1,1'-Biphenyl]-4-ylmethyl)-3-(2-phenoxyethoxy)aniline

Supplier: Matrix Scientific

032084-500MG
101832-992EA 306.15 USD
101832-992
N-([1,1'-Biphenyl]-4-ylmethyl)-3-(2-phenoxyethoxy)aniline
N-([1,1'-Biphenyl]-4-ylmethyl)-3-(2-phenoxyethoxy)aniline

Hazard: Irritant

Formula: C₂₇H₂₅NO₂
MDL Number: MFCD10688095
CAS Number: 1040685-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR