N-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]-3,4-dimethylaniline

Supplier: Matrix Scientific

032083-500MG
101832-990EA 306.15 USD
101832-990
N-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]-3,4-dimethylaniline
N-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]-3,4-dimethylaniline

Hazard: Irritant

Formula: C₁₈H₂₂ClNO
MDL Number: MFCD10688094
CAS Number: 1040685-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR