3-Ethoxy-N-(4-isopropoxybenzyl)aniline

Supplier: Matrix Scientific

032079-500MG
101832-982EA 260.23 USD
101832-982
3-Ethoxy-N-(4-isopropoxybenzyl)aniline
3-Ethoxy-N-(4-isopropoxybenzyl)aniline

CAS: 1040685-64-0
MDL Number: MFCD10688090
Molecular Formula: C18H23NO2
Molecular Weight: 285,39
Hazard: Irritant

Formula: C₁₈H₂₃NO₂
MW: 285,39 g/mol
MDL Number: MFCD10688090
CAS Number: 1040685-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR