N-(4-Butoxybenzyl)-3-ethoxyaniline

Supplier: Matrix Scientific

032077-500MG
101832-978EA 306.15 USD
101832-978
N-(4-Butoxybenzyl)-3-ethoxyaniline
N-(4-Butoxybenzyl)-3-ethoxyaniline

Hazard: Irritant

Formula: C₁₉H₂₅NO₂
MDL Number: MFCD10688088
CAS Number: 1040685-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR