3-Ethoxy-N-[2-(3-methylphenoxy)propyl]aniline

Supplier: Matrix Scientific

032074-500MG
101832-972EA 306.15 USD
101832-972
3-Ethoxy-N-[2-(3-methylphenoxy)propyl]aniline
3-Ethoxy-N-[2-(3-methylphenoxy)propyl]aniline

Hazard: Irritant

Formula: C₁₈H₂₃NO₂
MDL Number: MFCD10688085
CAS Number: 1040685-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR