3-Ethoxy-N-[2-(2-isopropyl-5-methylphenoxy)ethyl]aniline

Supplier: Matrix Scientific

032072-500MG
101832-968EA 306.15 USD
101832-968
3-Ethoxy-N-[2-(2-isopropyl-5-methylphenoxy)ethyl]aniline
3-Ethoxy-N-[2-(2-isopropyl-5-methylphenoxy)ethyl]aniline

Hazard: Irritant

Formula: C₂₀H₂₇NO₂
MDL Number: MFCD10688083
CAS Number: 1040685-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR