N-[4-(2,4-Dichlorophenoxy)butyl]-3-ethoxyaniline

Supplier: Matrix Scientific

032071-500MG
101832-966EA 306.15 USD
101832-966
N-[4-(2,4-Dichlorophenoxy)butyl]-3-ethoxyaniline
N-[4-(2,4-Dichlorophenoxy)butyl]-3-ethoxyaniline

Hazard: Irritant

Formula: C₁₈H₂₁Cl₂NO₂
MDL Number: MFCD10688082
CAS Number: 1040685-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR