N-[2-(4-Chloro-3-methylphenoxy)ethyl]-3-ethoxyaniline

Supplier: Matrix Scientific

032066-500MG
101832-956EA 306.15 USD
101832-956
N-[2-(4-Chloro-3-methylphenoxy)ethyl]-3-ethoxyaniline
N-[2-(4-Chloro-3-methylphenoxy)ethyl]-3-ethoxyaniline

Hazard: Irritant

Formula: C₁₇H₂₀ClNO₂
MDL Number: MFCD10688077
CAS Number: 1040684-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR