3-(Isopentyloxy)-N-(3-isopropoxybenzyl)aniline

Supplier: Matrix Scientific

032065-500MG
101832-954EA 306.15 USD
101832-954
3-(Isopentyloxy)-N-(3-isopropoxybenzyl)aniline
3-(Isopentyloxy)-N-(3-isopropoxybenzyl)aniline

Hazard: Irritant

Formula: C₂₁H₂₉NO₂
MDL Number: MFCD10688076
CAS Number: 1040684-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR