N-(2-Ethoxybenzyl)-3-(isopentyloxy)aniline

Supplier: Matrix Scientific

032061-500MG
101832-946EA 306.15 USD
101832-946
N-(2-Ethoxybenzyl)-3-(isopentyloxy)aniline
N-(2-Ethoxybenzyl)-3-(isopentyloxy)aniline

Hazard: Irritant

Formula: C₂₀H₂₇NO₂
MDL Number: MFCD10688072
CAS Number: 1040684-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR