N-[2-(2-Chlorophenoxy)ethyl]-3-(isopentyloxy)aniline

Supplier: Matrix Scientific

032060-500MG
101832-944EA 306.15 USD
101832-944
N-[2-(2-Chlorophenoxy)ethyl]-3-(isopentyloxy)aniline
N-[2-(2-Chlorophenoxy)ethyl]-3-(isopentyloxy)aniline

Hazard: Irritant

Formula: C₁₉H₂₄ClNO₂
MDL Number: MFCD10688071
CAS Number: 1040684-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR