3-(Isopentyloxy)-N-(2-thienylmethyl)aniline

Supplier: Matrix Scientific

032057-500MG
101832-938EA 306.15 USD
101832-938
3-(Isopentyloxy)-N-(2-thienylmethyl)aniline
3-(Isopentyloxy)-N-(2-thienylmethyl)aniline

Hazard: Irritant

Formula: C₁₆H₂₁NOS
MDL Number: MFCD10688068
CAS Number: 1040684-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR