N-(2-Fluorobenzyl)-3-propoxyaniline

Supplier: Matrix Scientific

032056-500MG
101832-936EA 306.15 USD
101832-936
N-(2-Fluorobenzyl)-3-propoxyaniline
N-(2-Fluorobenzyl)-3-propoxyaniline

Hazard: Irritant

Formula: C₁₆H₁₈FNO
MDL Number: MFCD10688067
CAS Number: 1040684-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR