N-(4-Butoxybenzyl)-3-propoxyaniline

Supplier: Matrix Scientific

032055-500MG
101832-934EA 306.15 USD
101832-934
N-(4-Butoxybenzyl)-3-propoxyaniline
N-(4-Butoxybenzyl)-3-propoxyaniline

Hazard: Irritant

Formula: C₂₀H₂₇NO₂
MDL Number: MFCD10688066
CAS Number: 1040684-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR