N-(4-Chlorophenethyl)-3-propoxyaniline

Supplier: Matrix Scientific

032054-500MG
101832-932EA 306.15 USD
101832-932
N-(4-Chlorophenethyl)-3-propoxyaniline
N-(4-Chlorophenethyl)-3-propoxyaniline

Hazard: Irritant

Formula: C₁₇H₂₀ClNO
MDL Number: MFCD10688065
CAS Number: 1040683-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR