N-[2-(2-Methylphenoxy)butyl]-3-propoxyaniline

Supplier: Matrix Scientific

032053-500MG
101832-930EA 306.15 USD
101832-930
N-[2-(2-Methylphenoxy)butyl]-3-propoxyaniline
N-[2-(2-Methylphenoxy)butyl]-3-propoxyaniline

Hazard: Irritant

Formula: C₂₀H₂₇NO₂
MDL Number: MFCD10688064
CAS Number: 1040683-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR