N-[2-(4-Methoxyphenoxy)ethyl]-3-propoxyaniline

Supplier: Matrix Scientific

032051-500MG
101832-926EA 306.15 USD
101832-926
N-[2-(4-Methoxyphenoxy)ethyl]-3-propoxyaniline
N-[2-(4-Methoxyphenoxy)ethyl]-3-propoxyaniline

Hazard: Irritant

Formula: C₁₈H₂₃NO₃
MDL Number: MFCD10688062
CAS Number: 1040683-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR