N-(3-Butoxyphenyl)-N-[4-(tetrahydro-2-furanylmethoxy)benzyl]amine

Supplier: Matrix Scientific

032049-500MG
101832-922EA 306.15 USD
101832-922
N-(3-Butoxyphenyl)-N-[4-(tetrahydro-2-furanylmethoxy)benzyl]amine
N-(3-Butoxyphenyl)-N-[4-(tetrahydro-2-furanylmethoxy)benzyl]amine

Hazard: Irritant

Formula: C₂₂H₂₉NO₃
MDL Number: MFCD10688060
CAS Number: 1040683-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR