3-Butoxy-N-[2-(4-ethylphenoxy)propyl]aniline

Supplier: Matrix Scientific

032046-500MG
101832-916EA 306.15 USD
101832-916
3-Butoxy-N-[2-(4-ethylphenoxy)propyl]aniline
3-Butoxy-N-[2-(4-ethylphenoxy)propyl]aniline

Hazard: Irritant

Formula: C₂₁H₂₉NO₂
MDL Number: MFCD10688057
CAS Number: 1040683-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR