4-Chloro-N-(4-propoxybenzyl)aniline

Supplier: Matrix Scientific

032036-500MG
101832-896EA 306.15 USD
101832-896
4-Chloro-N-(4-propoxybenzyl)aniline
4-Chloro-N-(4-propoxybenzyl)aniline

Hazard: Irritant

Formula: C₁₆H₁₈ClNO
MDL Number: MFCD04322321
CAS Number: 1036585-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR