4-Chloro-N-[4-(hexyloxy)benzyl]aniline

Supplier: Matrix Scientific

032034-500MG
101832-892EA 306.15 USD
101832-892
4-Chloro-N-[4-(hexyloxy)benzyl]aniline
4-Chloro-N-[4-(hexyloxy)benzyl]aniline

Hazard: Irritant

Formula: C₁₉H₂₄ClNO
MDL Number: MFCD10688046
CAS Number: 1040682-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR