3-(Hexyloxy)-N-(3-methylbenzyl)aniline

Supplier: Matrix Scientific

032032-500MG
101832-888EA 306.15 USD
101832-888
3-(Hexyloxy)-N-(3-methylbenzyl)aniline
3-(Hexyloxy)-N-(3-methylbenzyl)aniline

Hazard: Irritant

Formula: C₂₀H₂₇NO
MDL Number: MFCD10688044
CAS Number: 1040682-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR