2-Isopropyl-N-[4-(2-methoxyethoxy)benzyl]aniline

Supplier: Matrix Scientific

032028-500MG
101832-880EA 306.15 USD
101832-880
2-Isopropyl-N-[4-(2-methoxyethoxy)benzyl]aniline
2-Isopropyl-N-[4-(2-methoxyethoxy)benzyl]aniline

Hazard: Irritant

Formula: C₁₉H₂₅NO₂
MDL Number: MFCD10688040
CAS Number: 1040682-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR