2-Isopropyl-N-octylaniline

Supplier: Matrix Scientific

032027-500MG
101832-878EA 306.15 USD
101832-878
2-Isopropyl-N-octylaniline
2-Isopropyl-N-octylaniline

Hazard: Irritant

Formula: C₁₇H₂₉N
MDL Number: MFCD10688039
CAS Number: 1038724-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR