N-[2-(2-Chlorophenoxy)propyl]-2-isopropylaniline
Supplier: Matrix Scientific
032025-500MG
101832-874EA
306.15
USD
101832-874
N-[2-(2-Chlorophenoxy)propyl]-2-isopropylaniline
N-[2-(2-Chlorophenoxy)propyl]-2-isopropylaniline
Hazard: Irritant
Formula:
C₁₈H₂₂ClNO |
MDL Number:
MFCD10688037 CAS Number: 1040681-83-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...