N-(3-Chlorobenzyl)-2-isopropylaniline

Supplier: Matrix Scientific

032024-500MG
101832-872EA 306.15 USD
101832-872
N-(3-Chlorobenzyl)-2-isopropylaniline
N-(3-Chlorobenzyl)-2-isopropylaniline

Hazard: Irritant

Formula: C₁₆H₁₈ClN
MDL Number: MFCD10688036
CAS Number: 1039820-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR