2-Isopropyl-N-[2-(2-isopropyl-5-methylphenoxy)ethyl]aniline

Supplier: Matrix Scientific

032022-500MG
101832-868EA 306.15 USD
101832-868
2-Isopropyl-N-[2-(2-isopropyl-5-methylphenoxy)ethyl]aniline
2-Isopropyl-N-[2-(2-isopropyl-5-methylphenoxy)ethyl]aniline

Hazard: Irritant

Formula: C₂₁H₂₉NO
MDL Number: MFCD10688034
CAS Number: 1040681-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR